Gene Transcripts
- Gene Symbol
- Olfr292
- Entrez Gene ID
- 258613
- Gene Name
- olfactory receptor 292
- Gene Aliases
- MOR220-2
- Chromosome Location
- Chr. 7: 86688341 - 86695384 on Build GRCm38
- UniGene ID
- Mm.271466
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146620.2 |
NP_666831.2
|
2 |
480 |
|
View Full Product Details