Gene Transcripts

Gene Symbol
Olfr1273-ps
Entrez Gene ID
258977
Gene Name
olfactory receptor 1273, pseudogene
Gene Aliases
MOR227-4, Olfr1273
Chromosome Location
Chr. 2: 90295936 - 90296859 on Build GRCm38
UniGene ID
Mm.377774
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146975.1 NP_667186.1 1 193
GenBank mRNA BC104266.1 - 1 193
BC104267.1 - 1 193 40040403
BC126917.1 - 1 193 40040394

View Full Product Details