Gene Transcripts
- Gene Symbol
- Olfr584
- Entrez Gene ID
- 259056
- Gene Name
- olfactory receptor 584
- Gene Aliases
- MOR30-2
- Chromosome Location
- Chr. 7: 103085520 - 103086479 on Build GRCm38
- UniGene ID
- Mm.377813
- Species
- Mus musculus
- Gene Symbol
- Olfr582
- Entrez Gene ID
- 259055
- Gene Name
- olfactory receptor 582
- Gene Aliases
- MOR30-3
- Chromosome Location
- Chr. 7: 103041481 - 103042440 on Build GRCm38
- UniGene ID
- Mm.377812
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC107338.1 |
- |
1 |
43 |
|
BC107339.1 |
- |
1 |
43 |
|
View Full Product Details