Gene Transcripts

Gene Symbol
Sntg2
Entrez Gene ID
268534
Gene Name
syntrophin, gamma 2
Gene Aliases
2210008K22Rik, 9530013L23, BB121248
Chromosome Location
Chr. 12: 30174480 - 30373392 on Build GRCm38
UniGene ID
Mm.279765
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172951.3 NP_766539.2 7 600
GenBank mRNA AF367760.1 - 7 455
AK008673.1 - 7 508
AK035310.1 - 7 554
AK087560.1 - 7 550
AK137345.1 - 7 559
BC058204.1 - 7 490
BC138956.1 - 7 514 8861978
BC145476.1 - 7 509

View Full Product Details