Gene Transcripts

Gene Symbol
6330408A02Rik
Entrez Gene ID
321008
Gene Name
RIKEN cDNA 6330408A02 gene
Gene Aliases
Chromosome Location
Chr. 7: 13258967 - 13278721 on Build GRCm38
UniGene ID
Mm.458832
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177312.4 NP_796286.2 4 1203
GenBank mRNA AK018141.1 - 4 963
AK160064.1 - 3 1842
BC075681.1 - 3 1177 30608389

View Full Product Details