Gene Transcripts
- Gene Symbol
- Arl14epl
- Entrez Gene ID
- 381142
- Gene Name
- ADP-ribosylation factor-like 14 effector protein-like
- Gene Aliases
- Gm949
- Chromosome Location
- Chr. 18: 46921812 - 46934222 on Build GRCm38
- UniGene ID
- Mm.329423
- Species
- Mus musculus
- Gene Symbol
- Gm7904
- Entrez Gene ID
- 666043
- Gene Name
- predicted gene 7904
- Gene Aliases
- EG666043
- Chromosome Location
- Chr. 9: 21607077 - 21608329 on Build GRCm38
- UniGene ID
- Mm.247282
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC028798.1 |
- |
1 |
81 |
|
View Full Product Details