Gene Transcripts

Gene Symbol
Zfp69
Entrez Gene ID
381549
Gene Name
zinc finger protein 69
Gene Aliases
Gm1029, Krab2, Zfp63
Chromosome Location
Chr. 4: 120930133 - 120951701 on Build GRCm38
UniGene ID
Mm.331840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005788.3 NP_001005788.2 6 1132
GenBank mRNA BC066020.1 - 5 731
U29497.1 - 3 280

View Full Product Details