Gene Transcripts

Gene Symbol
1700123I01Rik
Entrez Gene ID
622554
Gene Name
RIKEN cDNA 1700123I01 gene
Gene Aliases
Chromosome Location
Chr. 19: 6175080 - 6225885 on Build GRCm38
UniGene ID
Mm.159757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165919.1 NP_001159391.1 3 305
GenBank mRNA AK007250.1 - 3 311
BC138005.1 - 3 219 8861024
BC145092.1 - 3 219 9053640

View Full Product Details