Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172253.2 | NP_758457.1 | 3 | 565 | |
GenBank mRNA | AK008677.1 | - | 3 | 547 | |
AK010829.1 | - | 3 | 566 | ||
AK016638.1 | - | 3 | 562 | ||
AK017016.1 | - | 3 | 555 | ||
AK017810.1 | - | 3 | 550 | ||
AK153090.1 | - | 555 | |||
AK160682.1 | - | 3 | 553 | ||
BC082580.1 | - | 537 | |||
BC139162.1 | - | 3 | 553 | 8862184 | |
BC139163.1 | - | 3 | 553 | 8862185 |