Gene Transcripts

Gene Symbol
Vmn2r110
Entrez Gene ID
224582
Gene Name
vomeronasal 2, receptor 110
Gene Aliases
EG224582
Chromosome Location
Chr. 17: 20573829 - 20596259 on Build GRCm38
UniGene ID
Mm.469851
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104572.1 NP_001098042.1 3 956

View Full Product Details