Gene Transcripts

Gene Symbol
Cyp2c50
Entrez Gene ID
107141
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 50
Gene Aliases
AI266900
Chromosome Location
Chr. 19: 40089679 - 40113955 on Build GRCm38
UniGene ID
Mm.474575
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167875.1 NP_001161347.1 2 325
NM_001167877.1 NP_001161349.1 2 325
NM_134144.2 NP_598905.2 2 325
GenBank mRNA AK149466.1 - 2 270
AY206873.1 - 2 304
BC011222.1 - 2 308 4195223
BC051050.1 - 2 316 5052536
BY086770.1 - 2 328

Gene Symbol
Cyp2c54
Entrez Gene ID
404195
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 54
Gene Aliases
EG404195
Chromosome Location
Chr. 19: 40037940 - 40073845 on Build GRCm38
UniGene ID
Mm.379575
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206537.2 NP_996260.1 2 306
GenBank mRNA AI181879.1 - 2 295
BC043929.1 - 1 88
BF659738.1 - 2 306

View Full Product Details