Gene Transcripts

Gene Symbol
Gm12171
Entrez Gene ID
675689
Gene Name
predicted gene 12171
Gene Aliases
OTTMUSG00000005499
Chromosome Location
Chr. 11: 46547131 - 46556358 on Build GRCm38
UniGene ID
Mm.478806
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163353.1 NP_001156825.1 2, 3 106

View Full Product Details