Gene Transcripts

Gene Symbol
Sod2
Entrez Gene ID
24787
Gene Name
superoxide dismutase 2, mitochondrial
Gene Aliases
Chromosome Location
Chr. 1: 47914757 - 47921587 on Build Rnor_6.0
UniGene ID
Rn.10488
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017051.2 NP_058747.1 5 652
GenBank mRNA BC070913.1 - 5 652
Y00497.1 - 5 580

View Full Product Details