Gene Transcripts

Gene Symbol
Apoa1
Entrez Gene ID
25081
Gene Name
apolipoprotein A-I
Gene Aliases
apoA-I
Chromosome Location
Chr. 8: 50525091 - 50526875 on Build Rnor_6.0
UniGene ID
Rn.10308
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012738.1 NP_036870.1 4 659
GenBank mRNA BC089820.1 - 3 643
FQ209434.1 - 4 660
FQ209676.1 - 4 664
FQ209761.1 - 4 660
FQ209776.1 - 5 615
FQ209818.1 - 4 665
FQ209900.1 - 4 660
FQ209928.1 - 4 664
FQ209958.1 - 4 664
FQ210010.1 - 4 664
FQ210126.1 - 4 661
FQ210161.1 - 4 687
FQ210244.1 - 1 321
FQ210354.1 - 4 664
FQ210443.1 - 4 661
FQ210470.1 - 4 660
FQ210544.1 - 4 662
FQ210653.1 - 4 662
FQ210722.1 - 4 664
FQ210732.1 - 1 301
FQ210743.1 - 4 767
FQ210863.1 - 4 664
FQ210880.1 - 4 660
FQ210914.1 - 4 660
FQ210920.1 - 4 660
FQ211297.1 - 4 662
FQ211535.1 - 4 667
FQ211571.1 - 4 660
FQ218078.1 - 4 660
FQ218079.1 - 4 664
FQ218325.1 - 4 664
FQ218519.1 - 4 660
FQ218525.1 - 4 660
FQ218615.1 - 4 659
FQ218659.1 - 4 663
FQ218663.1 - 4 660
FQ218837.1 - 4 666
FQ219021.1 - 4 664
FQ219052.1 - 4 666
FQ219074.1 - 4 660
FQ219479.1 - 4 752
FQ219515.1 - 4 664
FQ219577.1 - 4 660
FQ219728.1 - 4 659
FQ220986.1 - 4 661
FQ224655.1 - 4 660
FQ229884.1 - 4 699
M00001.1 - 4 659
X00558.1 - 3 648

View Full Product Details