Gene Transcripts
- Gene Symbol
- Th
- Entrez Gene ID
- 25085
- Gene Name
- tyrosine hydroxylase
- Gene Aliases
- The
- Chromosome Location
- Chr. 1: 216073034 - 216080287 on Build Rnor_6.0
- UniGene ID
- Rn.11082
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012740.3 |
NP_036872.1
|
5, 6 |
661 |
|
GenBank mRNA |
M10244.1 |
- |
5, 6 |
661 |
|
View Full Product Details