Gene Transcripts
- Gene Symbol
- Phex
- Entrez Gene ID
- 25512
- Gene Name
- phosphate regulating endopeptidase homolog, X-linked
- Gene Aliases
- PEX
- Chromosome Location
- Chr. X: 40460047 - 40717982 on Build Rnor_6.0
- UniGene ID
- Rn.37861
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013004.1 |
NP_037136.1
|
15 |
2090 |
|
GenBank mRNA |
AJ001637.1 |
- |
15 |
2090 |
|
View Full Product Details