Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017275.1 | NP_058971.1 | 9 | 965 | |
XM_006229572.2 | XP_006229634.1 | 9 | 797 | ||
XM_006229573.2 | XP_006229635.1 | 9 | 821 | ||
XM_006229574.2 | XP_006229636.1 | 9 | 771 | ||
XM_006229575.2 | XP_006229637.1 | 9 | 775 | ||
XM_008773628.1 | XP_008771850.1 | 9 | 890 | ||
XM_008773629.1 | XP_008771851.1 | 8 | 768 | ||
GenBank mRNA | AB004267.1 | - | 9 | 965 | |
D86556.1 | - | 9 | 816 |