Gene Transcripts

Gene Symbol
Ube2g1
Entrez Gene ID
64631
Gene Name
ubiquitin-conjugating enzyme E2G 1
Gene Aliases
Ubc7
Chromosome Location
Chr. 10: 59173268 - 59254458 on Build Rnor_6.0
UniGene ID
Rn.163149
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022690.2 NP_073181.1 2 402
GenBank mRNA AF099093.1 - 2 369
BC086980.1 - 2 402 7192141

View Full Product Details