Gene Transcripts

Gene Symbol
Lphn1
Entrez Gene ID
65096
Gene Name
latrophilin 1
Gene Aliases
CL1BA
Chromosome Location
Chr. 19: 25029037 - 25069146 on Build Rnor_6.0
UniGene ID
Rn.10776
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022962.1 NP_075251.1 11 2276
GenBank mRNA AF081144.1 - 10 2261
AF081145.1 - 10 2261
AF081146.1 - 11 2276
AF081147.1 - 11 2276
AF111099.1 - 12 2602
U72487.1 - 10 2078
U78105.1 - 10 2403

View Full Product Details