Gene Transcripts

Gene Symbol
Ido1
Entrez Gene ID
66029
Gene Name
indoleamine 2,3-dioxygenase 1
Gene Aliases
Ido, Indo
Chromosome Location
Chr. 16: 72216326 - 72228098 on Build Rnor_6.0
UniGene ID
Rn.64513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023973.1 NP_076463.1 1 46
GenBank mRNA AF312699.1 - 1 46

View Full Product Details