Gene Transcripts
- Gene Symbol
- Cdc42bpa
- Entrez Gene ID
- 114116
- Gene Name
- CDC42 binding protein kinase alpha
- Gene Aliases
- Mrck, Pk428
- Chromosome Location
- Chr. 13: 98231326 - 98447762 on Build Rnor_6.0
- UniGene ID
- Rn.10871
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053657.1 |
NP_446109.1
|
16 |
2823 |
|
GenBank mRNA |
AF021935.1 |
- |
16 |
2823 |
|
View Full Product Details