Gene Transcripts

Gene Symbol
Olr576
Entrez Gene ID
295814
Gene Name
olfactory receptor 576
Gene Aliases
Chromosome Location
Chr. 3: 75719373 - 75720299 on Build Rnor_6.0
UniGene ID
Rn.142896
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000328.1 NP_001000328.1 1 555

View Full Product Details