Gene Transcripts

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
Chromosome Location
Chr. 2: 97712030 - 97744327 on Build GRCh38
UniGene ID
Hs.234569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079.3 NP_001070.2 9 1214
NM_207519.1 NP_997402.1 1 232
XM_017004867.1 XP_016860356.1 11 2615
XM_017004868.1 XP_016860357.1 11 2597
XM_017004869.1 XP_016860358.1 11 2615
XM_017004870.1 XP_016860359.1 11 2615
GenBank mRNA AB083211.1 - 1 232
AL832287.1 - 3 1522
BC039039.1 - 8 868 5168073
BC053878.1 - 9 1190 5757161
L05148.1 - 9 1216

View Full Product Details