Gene Transcripts

Gene Symbol
Sntg2
Entrez Gene ID
298936
Gene Name
syntrophin, gamma 2
Gene Aliases
Chromosome Location
Chr. 6: 49123084 - 49324530 on Build Rnor_6.0
UniGene ID
Rn.212984
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239967.2 XP_006240029.1 3, 4 421
XM_006239968.2 XP_006240030.1 3, 4 421

View Full Product Details