Gene Transcripts

Gene Symbol
Ifi27l2b
Entrez Gene ID
299269
Gene Name
interferon, alpha-inducible protein 27 like 2B
Gene Aliases
isg12(b)
Chromosome Location
Chr. 6: 127336305 - 127337791 on Build Rnor_6.0
UniGene ID
Rn.22509
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206846.2 NP_996728.1 4 412
GenBank mRNA BC166870.1 - 4 412

View Full Product Details