Gene Transcripts

Gene Symbol
Olr1320
Entrez Gene ID
300621
Gene Name
olfactory receptor 1320
Gene Aliases
Chromosome Location
Chr. 8: 43628182 - 43629120 on Build Rnor_6.0
UniGene ID
Rn.143478
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000471.1 NP_001000471.1 1 474

View Full Product Details