Gene Transcripts

Gene Symbol
Polr1c
Entrez Gene ID
301246
Gene Name
polymerase (RNA) I polypeptide C
Gene Aliases
Ac2-127, Rpo1-1
Chromosome Location
Chr. 9: 17120759 - 17124871 on Build Rnor_6.0
UniGene ID
Rn.107990
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008330.1 NP_001008331.1 5, 6 566
GenBank mRNA AY325219.1 - 3, 4 292
BC086597.1 - 5, 6 566 7312277

View Full Product Details