Gene Transcripts

Gene Symbol
Mtif2
Entrez Gene ID
305606
Gene Name
mitochondrial translational initiation factor 2
Gene Aliases
Chromosome Location
Chr. 14: 113946084 - 113965574 on Build Rnor_6.0
UniGene ID
Rn.160632
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004254.1 NP_001004254.1 11 1640
GenBank mRNA BC079421.1 - 11 1640

View Full Product Details