Gene Transcripts

Gene Symbol
Olr1637
Entrez Gene ID
305841
Gene Name
olfactory receptor 1637
Gene Aliases
Chromosome Location
Chr. 15: 27996520 - 27999286 on Build Rnor_6.0
UniGene ID
Rn.142437
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000503.1 NP_001000503.1 1 822
XM_006251869.2 XP_006251931.1 2 879

View Full Product Details