Gene Transcripts

Gene Symbol
Mmrn2
Entrez Gene ID
306288
Gene Name
multimerin 2
Gene Aliases
Chromosome Location
Chr. 16: 10727497 - 10749307 on Build Rnor_6.0
UniGene ID
Rn.226152
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751550.3 XP_003751598.2 6 902
GenBank mRNA FQ218167.1 - 6 837

View Full Product Details