Gene Transcripts

Gene Symbol
Olfml3
Entrez Gene ID
310743
Gene Name
olfactomedin-like 3
Gene Aliases
Chromosome Location
Chr. 2: 206219419 - 206222248 on Build Rnor_6.0
UniGene ID
Rn.106761
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107708.1 NP_001101178.1 2 333
XM_006233077.1 XP_006233139.1 2 318
GenBank mRNA BC158838.1 - 2 272
FQ228335.1 - 2 285

View Full Product Details