Gene Transcripts

Gene Symbol
Myom3
Entrez Gene ID
313625
Gene Name
myomesin 3
Gene Aliases
Chromosome Location
Chr. 5: 154109289 - 154159943 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750060.3 XP_003750108.1 13 1630
XM_006239278.2 XP_006239340.1 13 1615

View Full Product Details