Gene Transcripts

Gene Symbol
Plekho2
Entrez Gene ID
315764
Gene Name
pleckstrin homology domain containing, family O member 2
Gene Aliases
Plekhq1, RGD1305718
Chromosome Location
Chr. 8: 71094272 - 71118968 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_236354.5 XP_236354.1 4, 5 520

View Full Product Details