Gene Transcripts

Gene Symbol
Col6a5
Entrez Gene ID
501047
Gene Name
collagen, type VI, alpha 5
Gene Aliases
RGD1565804
Chromosome Location
Chr. 8: 114527807 - 114632150 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766594.1 XP_008764816.1 4 1421

View Full Product Details