Gene Transcripts

Gene Symbol
Stx18
Entrez Gene ID
360953
Gene Name
syntaxin 18
Gene Aliases
Chromosome Location
Chr. 14: 77402952 - 77492910 on Build Rnor_6.0
UniGene ID
Rn.53006
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012151.1 NP_001012151.1 10 912
GenBank mRNA BC079183.1 - 10 912

View Full Product Details