Gene Transcripts

Gene Symbol
Galntl6
Entrez Gene ID
361142
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 6
Gene Aliases
Galnt17, RGD1564773
Chromosome Location
Chr. 16: 34553966 - 35803016 on Build Rnor_6.0
UniGene ID
Rn.214629
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135756.1 NP_001129228.1 9 1427
XM_008771227.1 XP_008769449.1 8 2081

View Full Product Details