Gene Transcripts

Gene Symbol
Ankzf1
Entrez Gene ID
363255
Gene Name
ankyrin repeat and zinc finger domain containing 1
Gene Aliases
RGD1359242
Chromosome Location
Chr. 9: 82393619 - 82400537 on Build Rnor_6.0
UniGene ID
Rn.163073
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014219.1 NP_001014241.1 7 1313
XM_006245248.2 XP_006245310.1 7 1366
GenBank mRNA BC081973.1 - 7 1313

View Full Product Details