Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021101.4 | NP_066924.1 | 4 | 819 | |
GenBank mRNA | AF101051.1 | - | 4 | 799 | |
AF114837.1 | - | 4 | 808 | ||
AF115546.1 | - | 4 | 589 | ||
AF134160.1 | - | 4 | 826 | ||
AK296934.1 | - | 5 | 700 | ||
AK312866.1 | - | 4 | 815 | ||
AY358652.1 | - | 4 | 797 | ||
BC012471.1 | - | 4 | 777 | 4500534 | |
CR457138.1 | - | 4 | 579 | ||
EF564137.1 | - | 4 | 579 |