Gene Transcripts

Gene Symbol
Olr551
Entrez Gene ID
404872
Gene Name
olfactory receptor 551
Gene Aliases
Chromosome Location
Chr. 3: 75045581 - 75045899 on Build Rnor_6.0
UniGene ID
Rn.142911
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000670.1 NP_001000670.1 44

View Full Product Details