Gene Transcripts

Gene Symbol
Olr14
Entrez Gene ID
404901
Gene Name
olfactory receptor 14
Gene Aliases
Chromosome Location
Chr. 1: 149816124 - 149817065 on Build Rnor_6.0
UniGene ID
Rn.141952
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000695.1 NP_001000695.1 1 245

Gene Symbol
LOC100910049
Entrez Gene ID
100910049
Gene Name
olfactory receptor 14A2-like
Gene Aliases
Chromosome Location
Chr. 1: 149630252 - 149633029 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748918.1 XP_003748966.1 245

View Full Product Details