Gene Transcripts
- Gene Symbol
- DYRK1B
- Entrez Gene ID
- 9149
- Gene Name
- dual specificity tyrosine phosphorylation regulated kinase 1B
- Gene Aliases
- AOMS3, MIRK
- Chromosome Location
- Chr. 19: 39825347 - 39834233 on Build GRCh38
- UniGene ID
- Hs.130988
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004714.2 |
NP_004705.1
|
3, 4 |
494 |
|
NM_006483.2 |
NP_006474.1
|
3, 4 |
494 |
|
NM_006484.2 |
NP_006475.1
|
3, 4 |
494 |
|
XM_005259398.4 |
XP_005259455.1
|
3, 4 |
946 |
|
GenBank mRNA |
AF205861.1 |
- |
3, 4 |
391 |
|
AK074971.1 |
- |
3, 4 |
410 |
|
AK302127.1 |
- |
3, 4 |
423 |
|
BC018751.1 |
- |
3, 4 |
446 |
4300012 |
BC025291.1 |
- |
3, 4 |
397 |
5019417 |
Y17999.1 |
- |
3, 4 |
462 |
|
View Full Product Details