Gene Transcripts

Gene Symbol
Zfp830
Entrez Gene ID
497967
Gene Name
zinc finger protein 830
Gene Aliases
Ccdc16, Znf830
Chromosome Location
Chr. 10: 70118250 - 70119714 on Build Rnor_6.0
UniGene ID
Rn.137587
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037360.1 NP_001032437.1 1 927
GenBank mRNA BC104717.1 - 1 927

View Full Product Details