Gene Transcripts
- Gene Symbol
- GSTO1
- Entrez Gene ID
- 9446
- Gene Name
- glutathione S-transferase omega 1
- Gene Aliases
- GSTO 1-1, GSTTLp28, HEL-S-21, P28, SPG-R
- Chromosome Location
- Chr. 10: 104254194 - 104267464 on Build GRCh38
- UniGene ID
- Hs.190028
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001191002.1 |
NP_001177931.1
|
4, 5 |
684 |
|
NM_001191003.1 |
NP_001177932.1
|
5, 6 |
836 |
|
NM_004832.2 |
NP_004823.1
|
5, 6 |
783 |
|
GenBank mRNA |
AF212303.1 |
- |
5, 6 |
590 |
|
AK313680.1 |
- |
5, 6 |
601 |
|
BC000127.1 |
- |
5, 6 |
593 |
3351359 |
BX537431.1 |
- |
5, 6 |
711 |
|
CF130513.1 |
- |
5, 6 |
560 |
|
CN283947.1 |
- |
4, 5 |
487 |
|
EU794619.1 |
- |
5, 6 |
590 |
|
U90313.1 |
- |
5, 6 |
573 |
|
View Full Product Details