Gene Transcripts

Gene Symbol
Tas2r108
Entrez Gene ID
554302
Gene Name
taste receptor, type 2, member 108
Gene Aliases
T2R16, Tas2r4
Chromosome Location
Chr. 4: 68656928 - 68657842 on Build Rnor_6.0
UniGene ID
Rn.154318
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024686.1 NP_001019857.1 1 713
GenBank mRNA AY916510.1 - 1 713

View Full Product Details