Gene Transcripts

Gene Symbol
Zfp397
Entrez Gene ID
100909408
Gene Name
zinc finger protein 397
Gene Aliases
Znf397
Chromosome Location
Chr. 18: 15856841 - 15864956 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271475.1 NP_001258404.1 2, 3 658
XM_006254456.2 XP_006254518.1 2, 3 675

View Full Product Details