Gene Transcripts

Gene Symbol
Krtap3-3l1
Entrez Gene ID
680060
Gene Name
keratin associated protein 3-3-like 1
Gene Aliases
Chromosome Location
Chr. 10: 87475253 - 87475869 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247444.2 XP_006247506.1 1 392

View Full Product Details