Gene Transcripts

Gene Symbol
Rbbp4
Entrez Gene ID
313048
Gene Name
retinoblastoma binding protein 4
Gene Aliases
Chromosome Location
Chr. 5: 147499880 - 147535240 on Build Rnor_6.0
UniGene ID
Rn.224577
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107912.2 NP_001101382.1 12 1760
GenBank mRNA BC168994.1 - 12 1760

View Full Product Details