Gene Transcripts

Gene Symbol
Prr19
Entrez Gene ID
681408
Gene Name
proline rich 19
Gene Aliases
Chromosome Location
Chr. 1: 82167037 - 82172636 on Build Rnor_6.0
UniGene ID
Rn.157169
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173428.1 NP_001166899.1 2 664
XM_006228478.2 XP_006228540.1 2 659
XM_006228479.2 XP_006228541.1 2 571
XM_006228480.2 XP_006228542.1 2 659
XM_008758985.1 XP_008757207.1 2 767
XM_008758986.1 XP_008757208.1 2 769
XM_008758987.1 XP_008757209.1 2 824
GenBank mRNA BC168703.1 - 2 662

View Full Product Details