Gene Transcripts

Gene Symbol
Polr3gl
Entrez Gene ID
690254
Gene Name
polymerase (RNA) III (DNA directed) polypeptide G-like
Gene Aliases
Chromosome Location
Chr. 2: 198698197 - 198719285 on Build Rnor_6.0
UniGene ID
Rn.86210
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109571.1 NP_001103041.1 8 710
GenBank mRNA BC168229.1 - 8 669
FQ214367.1 - 8 696

View Full Product Details