Gene Transcripts

Gene Symbol
Bpifb5
Entrez Gene ID
690479
Gene Name
BPI fold containing family B, member 5
Gene Aliases
Chromosome Location
Chr. 3: 149706004 - 149724469 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001074561.4 XP_001074561.2 4 558

View Full Product Details